The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007267.2 | NP_001007268.1 | 9 - 10 | 1760 | 90 |
NM_001195641.1 | NP_001182570.1 | 9 - 10 | 1760 | 90 | |
NM_007366.4 | NP_031392.3 | 9 - 10 | 1760 | 90 | |
XM_005246392.3 | XP_005246449.1 | 9 - 10 | 1648 | 90 | |
XM_011510820.2 | XP_011509122.1 | 9 - 10 | 1648 | 90 | |
XM_017003598.1 | XP_016859087.1 | 9 - 10 | 1648 | 90 | |
XM_017003599.1 | XP_016859088.1 | 9 - 10 | 1648 | 90 | |
XM_017003601.1 | XP_016859090.1 | 9 - 10 | 2047 | 90 | |
GenBank mRNA | BC140823.1 | - | 9 - 10 | 1570 | 90 |
BC144631.1 | - | 9 - 10 | 1570 | 90 | |
U17033.1 | - | 9 - 10 | 1760 | 90 | |
U17034.1 | - | 9 - 10 | 1760 | 90 |