The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 14 - 15 | 2127 | 116 |
NR_072994.1 | 14 - 15 | 2066 | 116 | ||
XM_006715130.2 | XP_006715193.1 | 14 - 15 | 2093 | 116 | |
XM_011514722.1 | XP_011513024.1 | 15 - 16 | 2117 | 116 | |
XM_017010995.1 | XP_016866484.1 | 13 - 14 | 2262 | 116 | |
GenBank mRNA | AJ544537.1 | - | 14 - 15 | 2060 | 116 |
BC136294.1 | - | 13 - 14 | 1711 | 116 | |
BC150310.1 | - | 14 - 15 | 2067 | 116 | |
Y13247.1 | - | 13 - 14 | 2050 | 116 |