The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 3 - 4 | 723 | 67 |
NR_072994.1 | 3 - 4 | 723 | 67 | ||
XM_006715130.2 | XP_006715193.1 | 3 - 4 | 689 | 67 | |
XM_011514722.1 | XP_011513024.1 | 4 - 5 | 713 | 67 | |
XM_017010995.1 | XP_016866484.1 | 2 - 3 | 858 | 67 | |
GenBank mRNA | AJ544537.1 | - | 3 - 4 | 656 | 67 |
AK307406.1 | - | 3 - 4 | 659 | 67 | |
BC005267.2 | - | 2 - 3 | 640 | 67 | |
BC136294.1 | - | 2 - 3 | 307 | 67 | |
BC150310.1 | - | 3 - 4 | 724 | 67 | |
Y13247.1 | - | 2 - 3 | 646 | 67 |