The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 9 - 10 | 1358 | 122 |
NR_072994.1 | 9 - 10 | 1358 | 122 | ||
XM_006715130.2 | XP_006715193.1 | 9 - 10 | 1324 | 122 | |
XM_011514722.1 | XP_011513024.1 | 10 - 11 | 1348 | 122 | |
XM_017010995.1 | XP_016866484.1 | 8 - 9 | 1493 | 122 | |
GenBank mRNA | AJ544537.1 | - | 9 - 10 | 1291 | 122 |
AK307406.1 | - | 9 - 10 | 1294 | 122 | |
BC005267.2 | - | 8 - 9 | 1275 | 122 | |
BC136294.1 | - | 8 - 9 | 942 | 122 | |
BC150310.1 | - | 9 - 10 | 1359 | 122 | |
Y13247.1 | - | 8 - 9 | 1281 | 122 |