The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005275.3 | NP_005266.2 | 9 - 10 | 2671 | 72 |
XM_005249015.3 | XP_005249072.1 | 10 - 11 | 2134 | 72 | |
GenBank mRNA | AK026391.1 | - | 9 - 10 | 1333 | 72 |
AK293596.1 | - | 3 - 4 | 624 | 72 | |
AK298042.1 | - | 9 - 10 | 1510 | 72 | |
AK301738.1 | - | 8 - 9 | 1066 | 72 | |
AK302480.1 | - | 8 - 9 | 1399 | 72 | |
AK316391.1 | - | 9 - 10 | 1272 | 72 | |
BC013959.2 | - | 9 - 10 | 1320 | 72 | |
BC018366.1 | - | 9 - 10 | 2251 | 72 | |
BC063657.1 | - | 6 - 7 | 903 | 72 | |
BT006648.1 | - | 9 - 10 | 1280 | 72 | |
L25665.1 | - | 9 - 10 | 72 | ||
X66436.1 | - | 1 - 2 | 72 |