The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281517.2 | NP_001268446.1 | 5 - 6 | 1181 | 74 |
NM_001319944.1 | NP_001306873.1 | 6 - 7 | 1334 | 74 | |
NM_022778.4 | NP_073615.2 | 6 - 7 | 1334 | 74 | |
XM_017002105.1 | XP_016857594.1 | 6 - 7 | 1334 | 74 | |
GenBank mRNA | AK024038.1 | - | 6 - 7 | 1290 | 74 |
AK299312.1 | - | 5 - 6 | 1134 | 74 | |
AL133609.1 | - | 2 - 3 | 175 | 74 | |
BC064528.1 | - | 6 - 7 | 1285 | 74 |