Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
Chromosome Location
Chr.7: 28995630 - 29146537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019029.2 NP_061902.2 2 - 3 369 114
NM_031311.3 NP_112601.3 2 - 3 281 114
XM_011515437.1 XP_011513739.1 2 - 3 448 114
XM_017012366.1 XP_016867855.1 2 - 3 281 114
GenBank mRNA AF106704.2 - 2 - 3 201 114
AF217508.1 - 1 - 2 167 114
AF282617.1 - 2 - 3 231 114
AK075433.1 - 2 - 3 282 114
AK124472.1 - 6 - 7 809 114
AK225545.1 - 2 - 3 232 114
AY358549.1 - 2 - 3 258 114
BC016838.2 - 2 - 3 329 114

View Full Product Details