The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019029.2 | NP_061902.2 | 6 - 7 | 755 | 102 |
NM_031311.3 | NP_112601.3 | 6 - 7 | 667 | 102 | |
XM_011515437.1 | XP_011513739.1 | 6 - 7 | 834 | 102 | |
XM_017012366.1 | XP_016867855.1 | 6 - 7 | 667 | 102 | |
GenBank mRNA | AF106704.2 | - | 6 - 7 | 587 | 102 |
AF217508.1 | - | 5 - 6 | 554 | 102 | |
AF282617.1 | - | 6 - 7 | 617 | 102 | |
AK075433.1 | - | 6 - 7 | 668 | 102 | |
AK124472.1 | - | 10 - 11 | 1195 | 102 | |
AK225545.1 | - | 6 - 7 | 618 | 102 | |
AY358549.1 | - | 6 - 7 | 644 | 102 | |
BC016838.2 | - | 6 - 7 | 715 | 102 |