The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019029.2 | NP_061902.2 | 9 - 10 | 1068 | 143 |
NM_031311.3 | NP_112601.3 | 9 - 10 | 980 | 143 | |
XM_011515437.1 | XP_011513739.1 | 9 - 10 | 1147 | 143 | |
GenBank mRNA | AF106704.2 | - | 9 - 10 | 143 | |
AF217508.1 | - | 8 - 9 | 867 | 143 | |
AF282617.1 | - | 9 - 10 | 143 | ||
AK124472.1 | - | 13 - 14 | 1508 | 143 | |
AK225545.1 | - | 9 - 10 | 931 | 143 | |
AY358549.1 | - | 9 - 10 | 957 | 143 | |
BC016838.2 | - | 9 - 10 | 1028 | 143 |