The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007033.1 | NP_001007034.1 | 4 - 5 | 485 | 72 |
NM_001317999.1 | NP_001304928.1 | 3 - 4 | 395 | 72 | |
GenBank mRNA | AK308011.1 | - | 4 - 5 | 553 | 72 |
AY321309.1 | - | 4 - 5 | 485 | 72 | |
AY365135.1 | - | 4 - 5 | 430 | 72 | |
BC132933.1 | - | 4 - 5 | 391 | 72 | |
BC132935.1 | - | 4 - 5 | 391 | 72 |