The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018009.3 | NP_001018009.2 | 8 - 9 | 1250 | 66 |
NM_004844.4 | NP_004835.2 | 8 - 9 | 1369 | 66 | |
XM_011534251.2 | XP_011532553.1 | 8 - 9 | 1181 | 66 | |
XM_017007521.1 | XP_016863010.1 | 8 - 9 | 1164 | 66 | |
XM_017007522.1 | XP_016863011.1 | 9 - 10 | 1807 | 66 | |
XM_017007523.1 | XP_016863012.1 | 8 - 9 | 1585 | 66 | |
XM_017007524.1 | XP_016863013.1 | 8 - 9 | 1117 | 66 | |
XM_017007525.1 | XP_016863014.1 | 8 - 9 | 1031 | 66 | |
GenBank mRNA | AB005047.1 | - | 8 - 9 | 1121 | 66 |
AK090524.1 | - | 10 - 11 | 1473 | 66 | |
AK313167.1 | - | 8 - 9 | 1327 | 66 | |
AL117422.1 | - | 8 - 9 | 1325 | 66 | |
BC006169.1 | - | 8 - 9 | 1224 | 66 | |
BC010123.2 | - | 8 - 9 | 1300 | 66 | |
BC023195.1 | - | 8 - 9 | 1041 | 66 | |
BX641159.1 | - | 1 - 2 | 1300 | 66 |