The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138809.3 | NP_620164.1 | 2 - 3 | 664 | 84 |
XM_017009034.1 | XP_016864523.1 | 2 - 3 | 428 | 84 | |
XM_017009035.1 | XP_016864524.1 | 2 - 3 | 384 | 84 | |
GenBank mRNA | AJ278125.1 | - | 2 - 3 | 457 | 84 |
AK074197.1 | - | 2 - 3 | 455 | 84 | |
AK315390.1 | - | 1 - 2 | 218 | 84 | |
BC001573.1 | - | 2 - 3 | 370 | 84 | |
BX647141.1 | - | 1 - 2 | 161 | 84 |