The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005691.3 | NP_005682.2 | 14 - 15 | 2041 | 64 |
NM_020297.3 | NP_064693.2 | 14 - 15 | 2041 | 64 | |
XM_005253284.3 | XP_005253341.1 | 16 - 17 | 2827 | 64 | |
XM_005253286.3 | XP_005253343.1 | 17 - 18 | 2887 | 64 | |
XM_005253287.4 | XP_005253344.1 | 16 - 17 | 2827 | 64 | |
XM_005253288.3 | XP_005253345.1 | 15 - 16 | 2711 | 64 | |
XM_005253289.3 | XP_005253346.1 | 16 - 17 | 2827 | 64 | |
XM_005253290.3 | XP_005253347.1 | 16 - 17 | 2827 | 64 | |
XM_006719025.3 | XP_006719088.1 | 16 - 17 | 2827 | 64 | |
XM_011520545.2 | XP_011518847.1 | 16 - 17 | 2883 | 64 |