The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002470.3 | NP_002461.2 | 19 - 20 | 2259 | 74 |
XM_011523870.2 | XP_011522172.1 | 19 - 20 | 3626 | 74 | |
XM_011523871.2 | XP_011522173.1 | 19 - 20 | 2259 | 74 |