The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001046.2 | NP_001037.1 | 18 - 19 | 2891 | 99 |
NM_001256461.1 | NP_001243390.1 | 18 - 19 | 2891 | 99 | |
NR_046207.1 | 18 - 19 | 2891 | 99 | ||
XM_017009771.1 | XP_016865260.1 | 17 - 18 | 2041 | 99 | |
GenBank mRNA | AF439152.1 | - | 5 - 6 | 512 | 99 |
AY280459.1 | - | 18 - 19 | 2916 | 99 | |
BC144221.1 | - | 18 - 19 | 2834 | 99 | |
BC146839.1 | - | 18 - 19 | 2834 | 99 | |
U30246.1 | - | 18 - 19 | 2891 | 99 |