Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC12A2
Entrez Gene ID
6558
Gene Name
solute carrier family 12 member 2
Gene Aliases
BSC, BSC2, NKCC1, PPP1R141
Chromosome Location
Chr.5: 128083791 - 128189688 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046.2 NP_001037.1 23 - 24 3381 130
NM_001256461.1 NP_001243390.1 22 - 23 3333 130
NR_046207.1 23 - 24 3381 130
XM_017009771.1 XP_016865260.1 22 - 23 2531 130
GenBank mRNA AY280459.1 - 23 - 24 3406 130
BC144221.1 - 22 - 23 3276 130
BC146839.1 - 22 - 23 3276 130
U30246.1 - 23 - 24 3381 130

View Full Product Details