The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001046.2 | NP_001037.1 | 6 - 7 | 1458 | 140 |
NM_001256461.1 | NP_001243390.1 | 6 - 7 | 1458 | 140 | |
NR_046207.1 | 6 - 7 | 1458 | 140 | ||
XM_011543588.2 | XP_011541890.1 | 6 - 7 | 1452 | 140 | |
XM_017009771.1 | XP_016865260.1 | 6 - 7 | 693 | 140 | |
GenBank mRNA | AY280459.1 | - | 6 - 7 | 1483 | 140 |
BC144221.1 | - | 6 - 7 | 1401 | 140 | |
BC146839.1 | - | 6 - 7 | 1401 | 140 | |
U30246.1 | - | 6 - 7 | 1458 | 140 |