Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISOC1
Entrez Gene ID
51015
Gene Name
isochorismatase domain containing 1
Gene Aliases
CGI-111
Chromosome Location
Chr.5: 129094749 - 129114028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016048.2 NP_057132.2 3 - 4 641 75
GenBank mRNA AF151869.1 - 3 - 4 640 75
AK025787.1 - 3 - 4 639 75
BC008367.1 - 3 - 4 619 75
BC014105.2 - 3 - 4 629 75

View Full Product Details