The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258039.1 | NP_001244968.1 | 1 - 2 | 125 | 122 |
XM_005262686.1 | XP_005262743.1 | 2 - 3 | 1042 | 122 | |
GenBank mRNA | AV692593.1 | - | 1 - 2 | 125 | 122 |
CN302060.1 | - | 1 - 2 | 105 | 122 |