Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3HC1
Entrez Gene ID
51530
Gene Name
zinc finger C3HC-type containing 1
Gene Aliases
HSPC216, NIPA
Chromosome Location
Chr.7: 130018286 - 130051451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282190.1 NP_001269119.1 1 - 2 236 88
GenBank mRNA AK001317.1 - 1 - 2 159 88

View Full Product Details