The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320766.1 | NP_001307695.1 | 19 - 20 | 3732 | 150 |
NM_015565.2 | NP_056380.2 | 20 - 21 | 3771 | 150 | |
XM_017028316.1 | XP_016883805.1 | 19 - 20 | 3451 | 150 | |
XM_017028317.1 | XP_016883806.1 | 10 - 11 | 2046 | 150 | |
GenBank mRNA | AB018257.2 | - | 20 - 21 | 3662 | 150 |
BC140790.1 | - | 20 - 21 | 3684 | 150 | |
BC150284.1 | - | 20 - 21 | 3662 | 150 | |
CN361592.1 | - | 4 - 5 | 434 | 150 |