Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
Chromosome Location
Chr.18: 9136753 - 9285985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083625.2 NP_001077094.1 2 - 3 350 121
NM_001204056.1 NP_001190985.1 2 - 3 448 121
NM_015208.4 NP_056023.3 2 - 3 350 121
XM_005258092.3 XP_005258149.1 2 - 3 483 121
XM_005258093.3 XP_005258150.1 2 - 3 467 121
XM_005258095.2 XP_005258152.1 2 - 3 322 121
XM_005258096.2 XP_005258153.1 2 - 3 326 121
XM_011525638.2 XP_011523940.1 2 - 3 322 121
XM_011525639.2 XP_011523941.1 2 - 3 439 121
XM_017025661.1 XP_016881150.1 2 - 3 451 121
XM_017025662.1 XP_016881151.1 2 - 3 496 121
XM_017025663.1 XP_016881152.1 2 - 3 435 121
XM_017025664.1 XP_016881153.1 2 - 3 435 121
XM_017025665.1 XP_016881154.1 2 - 3 469 121
XM_017025666.1 XP_016881155.1 2 - 3 408 121
XM_017025667.1 XP_016881156.1 2 - 3 264 121
XM_017025668.1 XP_016881157.1 2 - 3 264 121
GenBank mRNA AF317425.1 - 1 - 2 86 121
AK024808.1 - 2 - 3 300 121
AL834204.1 - 2 - 3 278 121
AY373757.1 - 2 - 3 300 121
AY533564.2 - 2 - 3 448 121
BC057225.1 - 2 - 3 299 121
BC070234.1 - 2 - 3 320 121
BP230422.1 - 2 - 3 350 121
BP255733.1 - 2 - 3 343 121

View Full Product Details