Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
Chromosome Location
Chr.16: 10944488 - 11193272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015226.2 NP_056041.1 22 - 23 2868 73
XM_005255210.1 XP_005255267.1 21 - 22 2856 73
XM_005255211.1 XP_005255268.1 22 - 23 2814 73
XM_005255213.1 XP_005255270.1 21 - 22 2808 73
XM_011522434.1 XP_011520736.1 21 - 22 2733 73
XM_011522435.1 XP_011520737.1 22 - 23 2862 73
XM_017023090.1 XP_016878579.1 10 - 11 1147 73
GenBank mRNA AB002348.3 - 22 - 23 2771 73
AK126771.1 - 1 - 2 76 73

View Full Product Details