Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D13
Entrez Gene ID
54662
Gene Name
TBC1 domain family member 13
Gene Aliases
-
Chromosome Location
Chr.9: 128787207 - 128810432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286772.1 NP_001273701.1 8 - 9 852 81
NM_018201.4 NP_060671.3 10 - 11 1227 81
XM_005252060.2 XP_005252117.1 9 - 10 1130 81
GenBank mRNA AB209565.1 - 7 - 8 946 81
AB449899.1 - 10 - 11 1080 81
AK001605.1 - 8 - 9 710 81
AK027457.1 - 2 - 3 271 81
AK123828.1 - 6 - 7 841 81
AK289777.1 - 10 - 11 1139 81
AK295183.1 - 7 - 8 930 81
AK307556.1 - 6 - 7 829 81
BC014898.1 - 10 - 11 1102 81
BC132817.1 - 10 - 11 1100 81
BC136509.1 - 10 - 11 1100 81
BC150311.1 - 10 - 11 1136 81

View Full Product Details