Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D13
Entrez Gene ID
54662
Gene Name
TBC1 domain family member 13
Gene Aliases
-
Chromosome Location
Chr.9: 128787207 - 128810432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286772.1 NP_001273701.1 1 - 2 171 72
NM_018201.4 NP_060671.3 1 - 2 171 72
XM_005252060.2 XP_005252117.1 1 - 2 115 72
GenBank mRNA AK123828.1 - 1 - 2 71 72
AK289777.1 - 1 - 2 83 72
BC014898.1 - 1 - 2 46 72
BC132817.1 - 1 - 2 44 72
BC136509.1 - 1 - 2 44 72
BC150311.1 - 1 - 2 80 72
BG716911.1 - 1 - 2 181 72
DC325060.1 - 1 - 2 147 72

View Full Product Details