Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D13
Entrez Gene ID
54662
Gene Name
TBC1 domain family member 13
Gene Aliases
-
Chromosome Location
Chr.9: 128787207 - 128810432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018201.4 NP_060671.3 8 - 9 906 70
XM_005252060.2 XP_005252117.1 7 - 8 809 70
GenBank mRNA AB209565.1 - 5 - 6 625 70
AB449899.1 - 8 - 9 759 70
AK123828.1 - 4 - 5 520 70
AK289777.1 - 8 - 9 818 70
AK295183.1 - 5 - 6 609 70
AK307556.1 - 4 - 5 508 70
BC014898.1 - 8 - 9 781 70
BC132817.1 - 8 - 9 779 70
BC136509.1 - 8 - 9 779 70
BC150311.1 - 8 - 9 815 70

View Full Product Details