Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL7
Entrez Gene ID
55975
Gene Name
kelch like family member 7
Gene Aliases
KLHL6, SBBI26
Chromosome Location
Chr.7: 23105734 - 23175421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031710.2 NP_001026880.2 7 - 8 1232 95
NM_018846.4 NP_061334.4 7 - 8 1316 95
NR_033328.1 8 - 9 1363 95
XM_006715753.2 XP_006715816.1 8 - 9 1266 95
XM_006715754.2 XP_006715817.1 9 - 10 1349 95
XM_006715755.2 XP_006715818.1 9 - 10 1568 95
XM_006715756.2 XP_006715819.1 8 - 9 1437 95
XM_017012439.1 XP_016867928.1 8 - 9 1529 95
GenBank mRNA AK056390.1 - 8 - 9 1314 95
AK290479.1 - 7 - 8 1026 95
AK296219.1 - 4 - 5 835 95
AK297369.1 - - 1055 95
AK297421.1 - 4 - 5 756 95
AK297595.1 - 6 - 7 1073 95
AK299006.1 - 7 - 8 958 95
AL136597.1 - 7 - 8 1178 95
AY028802.1 - 7 - 8 881 95
BC039585.1 - 7 - 8 1174 95
EF560731.1 - 8 - 9 1082 95

View Full Product Details