The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013628.2 | NP_001013650.1 | 1 - 1 | 879 | 72 |
GenBank mRNA | BC136673.1 | - | 2 - 2 | 787 | 72 |
BC136674.1 | - | 1 - 1 | 879 | 72 |