The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261406.1 | NP_001248335.1 | 30 - 31 | 4511 | 95 |
NM_001261407.1 | NP_001248336.1 | 30 - 31 | 4511 | 95 | |
NM_003829.4 | NP_003820.2 | 31 - 32 | 4610 | 95 | |
XM_005251622.4 | XP_005251679.1 | 31 - 32 | 4559 | 95 | |
XM_005251623.4 | XP_005251680.1 | 29 - 30 | 4349 | 95 | |
XM_006716885.3 | XP_006716948.1 | 31 - 32 | 4608 | 95 | |
XM_006716886.3 | XP_006716949.1 | 32 - 33 | 4767 | 95 | |
XM_006716887.3 | XP_006716950.1 | 31 - 32 | 4711 | 95 | |
XM_006716888.3 | XP_006716951.1 | 31 - 32 | 4559 | 95 | |
XM_006716889.3 | XP_006716952.1 | 30 - 31 | 4463 | 95 | |
XM_006716891.3 | XP_006716954.1 | 30 - 31 | 4463 | 95 | |
XM_017015252.1 | XP_016870741.1 | 31 - 32 | 4559 | 95 | |
XM_017015253.1 | XP_016870742.1 | 30 - 31 | 4448 | 95 | |
XM_017015254.1 | XP_016870743.1 | 30 - 31 | 4560 | 95 | |
XM_017015255.1 | XP_016870744.1 | 29 - 30 | 4408 | 95 | |
XM_017015256.1 | XP_016870745.1 | 29 - 30 | 4349 | 95 | |
XM_017015257.1 | XP_016870746.1 | 29 - 30 | 4336 | 95 | |
GenBank mRNA | AB210041.1 | - | 31 - 32 | 4611 | 95 |
AF093419.1 | - | 30 - 31 | 4434 | 95 | |
AK074721.1 | - | 1 - 2 | 95 | 95 | |
AK091945.1 | - | 6 - 7 | 888 | 95 | |
AK294823.1 | - | 4 - 5 | 520 | 95 | |
AK294976.1 | - | 9 - 10 | 1116 | 95 | |
AK307924.1 | - | 4 - 5 | 520 | 95 | |
AK307988.1 | - | 7 - 8 | 928 | 95 | |
AK316489.1 | - | 8 - 9 | 994 | 95 | |
BC140793.1 | - | 29 - 30 | 4335 | 95 | |
BC144564.1 | - | 29 - 30 | 4335 | 95 |