The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014429.3 | NP_055244.3 | 16 - 17 | 1650 | 134 |
XM_005247362.2 | XP_005247419.1 | 17 - 18 | 1697 | 134 | |
XM_011512691.1 | XP_011510993.1 | 17 - 18 | 1703 | 134 | |
XM_011512692.1 | XP_011510994.1 | 16 - 17 | 1613 | 134 | |
XM_011512693.1 | XP_011510995.1 | 17 - 18 | 1703 | 134 | |
XM_011512694.1 | XP_011510996.1 | 14 - 15 | 1454 | 134 | |
XM_011512696.2 | XP_011510998.1 | 11 - 12 | 1154 | 134 | |
XM_011512697.2 | XP_011510999.1 | 10 - 11 | 1322 | 134 | |
XM_017006169.1 | XP_016861658.1 | 17 - 18 | 1703 | 134 | |
XM_017006170.1 | XP_016861659.1 | 9 - 10 | 1032 | 134 | |
GenBank mRNA | AF084946.1 | - | 16 - 17 | 1625 | 134 |
AK302642.1 | - | 16 - 17 | 1607 | 134 | |
AL353955.1 | - | 10 - 11 | 1010 | 134 | |
BC028680.1 | - | 16 - 17 | 1651 | 134 | |
BC050307.1 | - | 16 - 17 | 1589 | 134 |