The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000053.3 | NP_000044.2 | 1 - 2 | 206 | 66 |
NM_001005918.2 | NP_001005918.1 | 1 - 2 | 206 | 66 | |
NM_001243182.1 | NP_001230111.1 | 1 - 2 | 206 | 66 | |
XM_005266427.2 | XP_005266484.1 | 1 - 2 | 268 | 66 | |
XM_005266428.1 | XP_005266485.1 | 1 - 2 | 268 | 66 | |
XM_005266430.4 | XP_005266487.1 | 2 - 3 | 214 | 66 | |
XM_005266432.2 | XP_005266489.1 | 1 - 2 | 268 | 66 | |
XM_011535118.1 | XP_011533420.1 | 1 - 2 | 268 | 66 | |
XM_011535119.1 | XP_011533421.1 | 1 - 2 | 268 | 66 | |
GenBank mRNA | BC117200.1 | - | 1 - 2 | 123 | 66 |
BC143973.1 | - | 1 - 2 | 123 | 66 | |
BC143974.1 | - | 1 - 2 | 121 | 66 | |
BC143975.1 | - | 1 - 2 | 123 | 66 | |
BC143976.1 | - | 1 - 2 | 123 | 66 | |
DA125470.1 | - | 1 - 2 | 268 | 66 | |
DQ015922.1 | - | 1 - 2 | 49 | 66 | |
U03464.1 | - | 1 - 2 | 49 | 66 | |
U11700.1 | - | 1 - 2 | 211 | 66 |