The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005898.4 | NP_005889.3 | 2 - 3 | 404 | 75 |
NM_203364.2 | NP_976240.1 | 2 - 3 | 404 | 75 | |
XM_017017749.1 | XP_016873238.1 | 1 - 2 | 375 | 75 | |
GenBank mRNA | BC001731.1 | - | 2 - 3 | 404 | 75 |
Z48042.1 | - | 1 - 2 | 257 | 75 |