The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320595.1 | NP_001307524.1 | 12 - 13 | 1643 | 134 |
NM_001320596.1 | NP_001307525.1 | 12 - 13 | 1532 | 134 | |
NM_001320597.1 | NP_001307526.1 | 11 - 12 | 1383 | 134 | |
NM_004396.4 | NP_004387.1 | 11 - 12 | 1618 | 134 | |
GenBank mRNA | AB451257.1 | - | 11 - 12 | 1216 | 134 |
AB451382.1 | - | 11 - 12 | 1216 | 134 | |
AK297192.1 | - | 10 - 11 | 1148 | 134 | |
AK297753.1 | - | 12 - 13 | 1353 | 134 | |
AK297905.1 | - | 10 - 11 | 1176 | 134 | |
AK310318.1 | - | 11 - 12 | 1262 | 134 | |
BC016027.1 | - | 11 - 12 | 1357 | 134 | |
BT006943.1 | - | 11 - 12 | 1216 | 134 | |
X15729.1 | - | 11 - 12 | 1383 | 134 | |
X52104.1 | - | 11 - 12 | 1391 | 134 |