The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003369.3 | NP_003360.2 | 10 - 11 | 1238 | 83 |
XM_011545226.2 | XP_011543528.1 | 10 - 11 | 1553 | 83 | |
XM_011545227.2 | XP_011543529.1 | 10 - 11 | 1553 | 83 | |
XM_011545228.2 | XP_011543530.1 | 10 - 11 | 1553 | 83 | |
XM_017018224.1 | XP_016873713.1 | 8 - 9 | 992 | 83 | |
XM_017018225.1 | XP_016873714.1 | 8 - 9 | 992 | 83 | |
XM_017018226.1 | XP_016873715.1 | 10 - 11 | 1553 | 83 | |
XM_017018227.1 | XP_016873716.1 | 4 - 5 | 403 | 83 | |
GenBank mRNA | AB012958.1 | - | 10 - 11 | 1175 | 83 |
AK095352.1 | - | 4 - 5 | 815 | 83 | |
AK222772.1 | - | 10 - 11 | 1129 | 83 | |
AK291011.1 | - | 10 - 11 | 1216 | 83 | |
AK293782.1 | - | 10 - 11 | 979 | 83 | |
AK296871.1 | - | 4 - 5 | 358 | 83 | |
AK311341.1 | - | 5 - 6 | 456 | 83 | |
AK316133.1 | - | 3 - 4 | 249 | 83 | |
BC064837.1 | - | 10 - 11 | 1148 | 83 |