The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022163.3 | NP_071446.2 | 3 - 4 | 611 | 79 |
GenBank mRNA | AF210056.1 | - | 3 - 4 | 597 | 79 |
AK315434.1 | - | 3 - 4 | 596 | 79 | |
BC017883.1 | - | 3 - 4 | 597 | 79 |