The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039690.3 | NP_001034779.1 | 2 - 3 | 165 | 75 |
NM_001040146.3 | NP_001035236.1 | 2 - 3 | 138 | 75 | |
XM_017023370.1 | XP_016878859.1 | 2 - 3 | 160 | 75 | |
GenBank mRNA | AF525164.1 | - | 2 - 3 | 145 | 75 |
AK000407.1 | - | 2 - 3 | 145 | 75 | |
BC018700.2 | - | 2 - 3 | 112 | 75 | |
BP222147.1 | - | 2 - 3 | 165 | 75 | |
CB135653.1 | - | 2 - 3 | 153 | 75 | |
CN388626.1 | - | 2 - 3 | 131 | 75 | |
DA076128.1 | - | 2 - 3 | 156 | 75 | |
DB188088.1 | - | 2 - 3 | 145 | 75 |