Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP4
Entrez Gene ID
84916
Gene Name
UTP4, small subunit processome component
Gene Aliases
CIRH1A, CIRHIN, NAIC, TEX292
Chromosome Location
Chr.16: 69132596 - 69169034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318391.1 NP_001305320.1 8 - 9 1045 87
NM_032830.2 NP_116219.2 8 - 9 1099 87
XM_005256205.3 XP_005256262.2 4 - 5 630 87
GenBank mRNA AB075868.2 - 8 - 9 1037 87
AK027419.1 - 5 - 6 654 87
AK027445.1 - 6 - 7 722 87
AK027634.1 - 8 - 9 1051 87
AK027675.1 - 8 - 9 1048 87
AK074795.1 - 8 - 9 1065 87
AK310104.1 - 8 - 9 87
BC000167.2 - 4 - 5 544 87
BC009348.2 - 8 - 9 1031 87
BX647265.1 - 8 - 9 1013 87

View Full Product Details