The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127384.2 | NP_001120856.1 | 11 - 12 | 1687 | 104 |
NM_013266.3 | NP_037398.2 | 11 - 12 | 1734 | 104 | |
XM_017016151.1 | XP_016871640.1 | 12 - 13 | 1729 | 104 | |
XM_017016152.1 | XP_016871641.1 | 14 - 15 | 2026 | 104 | |
XM_017016153.1 | XP_016871642.1 | 12 - 13 | 1737 | 104 | |
XM_017016154.1 | XP_016871643.1 | 7 - 8 | 1105 | 104 | |
XM_017016155.1 | XP_016871644.1 | 7 - 8 | 1115 | 104 | |
XM_017016156.1 | XP_016871645.1 | 7 - 8 | 992 | 104 | |
XM_017016157.1 | XP_016871646.1 | 5 - 6 | 1142 | 104 | |
GenBank mRNA | AF091606.1 | - | 11 - 12 | 1712 | 104 |
AK289756.1 | - | 11 - 12 | 1623 | 104 | |
AK313882.1 | - | 11 - 12 | 1737 | 104 |