The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122837.1 | NP_001116309.1 | 4 - 5 | 547 | 67 |
NM_001122839.1 | NP_001116311.1 | 4 - 5 | 509 | 67 | |
NM_001287586.1 | NP_001274515.1 | 4 - 5 | 574 | 67 | |
NM_001287587.1 | NP_001274516.1 | 5 - 6 | 691 | 67 | |
NM_001287589.1 | NP_001274518.1 | 3 - 4 | 385 | 67 | |
NM_001287590.1 | NP_001274519.1 | 4 - 5 | 704 | 67 | |
NM_001287591.1 | NP_001274520.1 | 5 - 6 | 728 | 67 | |
NM_001287592.1 | NP_001274521.1 | 3 - 4 | 460 | 67 | |
NM_018845.3 | NP_061333.2 | 5 - 6 | 671 | 67 | |
XM_005245339.4 | XP_005245396.1 | 5 - 6 | 857 | 67 | |
XM_005245340.3 | XP_005245397.1 | 5 - 6 | 938 | 67 | |
XM_005245341.4 | XP_005245398.1 | 5 - 6 | 889 | 67 | |
XM_005245342.3 | XP_005245399.1 | 4 - 5 | 497 | 67 | |
GenBank mRNA | AF126023.1 | - | 5 - 6 | 631 | 67 |
AF126024.1 | - | 4 - 5 | 496 | 67 | |
AK289551.1 | - | 5 - 6 | 672 | 67 | |
BC005943.1 | - | 5 - 6 | 615 | 67 | |
BC009621.1 | - | 4 - 5 | 471 | 67 | |
BC010869.1 | - | 4 - 5 | 462 | 67 | |
BC012378.1 | - | 4 - 5 | 496 | 67 | |
BI195885.1 | - | 4 - 5 | 706 | 67 | |
BM806886.1 | - | 5 - 6 | 715 | 67 | |
BU160680.1 | - | 5 - 6 | 613 | 67 | |
BX093804.1 | - | 3 - 4 | 377 | 67 | |
CR457148.1 | - | 5 - 6 | 571 | 67 | |
X84712.1 | - | 3 - 4 | 256 | 67 |