Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FERMT3
Entrez Gene ID
83706
Gene Name
fermitin family member 3
Gene Aliases
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF
Chromosome Location
Chr.11: 64206665 - 64223891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031471.5 NP_113659.3 8 - 9 1178 79
NM_178443.2 NP_848537.1 8 - 9 1178 79
XM_011545294.2 XP_011543596.1 8 - 9 1190 79
XM_011545295.2 XP_011543597.1 7 - 8 1023 79
XM_017018398.1 XP_016873887.1 8 - 9 1186 79
XM_017018399.1 XP_016873888.1 7 - 8 1038 79
GenBank mRNA AK093719.1 - 8 - 9 1123 79
AY093951.1 - 8 - 9 1124 79
AY093952.1 - 8 - 9 1124 79
BC004347.1 - 8 - 9 1122 79
BC013366.2 - 8 - 9 1122 79
BC015584.2 - 8 - 9 1117 79

View Full Product Details