The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031471.5 | NP_113659.3 | 9 - 10 | 1235 | 61 |
XM_017018398.1 | XP_016873887.1 | 9 - 10 | 1243 | 61 | |
XM_017018399.1 | XP_016873888.1 | 8 - 9 | 1095 | 61 | |
GenBank mRNA | AK093719.1 | - | 9 - 10 | 1180 | 61 |
AY093951.1 | - | 9 - 10 | 1181 | 61 | |
BC004347.1 | - | 9 - 10 | 1179 | 61 | |
BC013366.2 | - | 9 - 10 | 1179 | 61 | |
BC015584.2 | - | 9 - 10 | 1174 | 61 |