Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KSR1
Entrez Gene ID
8844
Gene Name
kinase suppressor of ras 1
Gene Aliases
KSR, RSU2
Chromosome Location
Chr.17: 27456441 - 27626438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014238.1 NP_055053.1 20 - 21 2590 88
XM_006722154.3 XP_006722217.1 17 - 18 2634 88
XM_011525429.2 XP_011523731.1 19 - 20 2742 88
XM_011525430.2 XP_011523732.1 19 - 20 2744 88
XM_011525431.2 XP_011523733.1 19 - 20 2744 88
XM_017025267.1 XP_016880756.1 18 - 19 2700 88
XM_017025268.1 XP_016880757.1 18 - 19 2676 88
XM_017025269.1 XP_016880758.1 18 - 19 2676 88
XM_017025270.1 XP_016880759.1 18 - 19 2477 88
XM_017025271.1 XP_016880760.1 20 - 21 3449 88
XM_017025272.1 XP_016880761.1 20 - 21 3432 88
XM_017025273.1 XP_016880762.1 19 - 20 3255 88
XM_017025274.1 XP_016880763.1 19 - 20 2458 88
XM_017025275.1 XP_016880764.1 19 - 20 2445 88
XM_017025276.1 XP_016880765.1 20 - 21 3397 88
XM_017025277.1 XP_016880766.1 15 - 16 2140 88
XM_017025278.1 XP_016880767.1 15 - 16 2150 88
GenBank mRNA U43586.1 - 15 - 16 1764 88

View Full Product Details