The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006722154.3 | XP_006722217.1 | 1 - 2 | 418 | 70 |
XM_011525429.2 | XP_011523731.1 | 1 - 2 | 418 | 70 | |
XM_011525430.2 | XP_011523732.1 | 1 - 2 | 420 | 70 | |
XM_011525431.2 | XP_011523733.1 | 1 - 2 | 420 | 70 | |
XM_017025267.1 | XP_016880756.1 | 1 - 2 | 418 | 70 | |
XM_017025268.1 | XP_016880757.1 | 1 - 2 | 418 | 70 | |
XM_017025269.1 | XP_016880758.1 | 1 - 2 | 418 | 70 | |
XM_017025279.1 | XP_016880768.1 | 1 - 2 | 435 | 70 | |
XM_017025280.1 | XP_016880769.1 | 1 - 2 | 435 | 70 |