Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCCC2
Entrez Gene ID
64087
Gene Name
methylcrotonoyl-CoA carboxylase 2
Gene Aliases
MCCB
Chromosome Location
Chr.5: 71587288 - 71658706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022132.4 NP_071415.1 11 - 12 1207 151
XM_005248567.1 XP_005248624.1 10 - 11 995 151
GenBank mRNA AB050049.1 - 11 - 12 1168 151
AF261884.1 - 11 - 12 1142 151
AF301000.1 - 11 - 12 1168 151
AF310971.1 - 11 - 12 1189 151
BC065027.1 - 11 - 12 1127 151

View Full Product Details