Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCCC2
Entrez Gene ID
64087
Gene Name
methylcrotonoyl-CoA carboxylase 2
Gene Aliases
MCCB
Chromosome Location
Chr.5: 71587288 - 71658706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022132.4 NP_071415.1 13 - 14 1351 97
XM_005248567.1 XP_005248624.1 12 - 13 1139 97
GenBank mRNA AB050049.1 - 13 - 14 1312 97
AF261884.1 - 13 - 14 1286 97
AF301000.1 - 13 - 14 1312 97
AF310971.1 - 13 - 14 1333 97
BC014897.1 - 12 - 13 1293 97
BC065027.1 - 13 - 14 1271 97
CN309723.1 - 1 - 2 62 97

View Full Product Details