The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000610.3 | NP_000601.3 | 17 - 18 | 2462 | 70 |
NM_001001389.1 | NP_001001389.1 | 16 - 17 | 2333 | 70 | |
NM_001001390.1 | NP_001001390.1 | 11 - 12 | 1715 | 70 | |
NM_001001391.1 | NP_001001391.1 | 8 - 9 | 1319 | 70 | |
NM_001001392.1 | NP_001001392.1 | 3 - 4 | 699 | 70 | |
NM_001202555.1 | NP_001189484.1 | 9 - 10 | 1523 | 70 | |
NM_001202556.1 | NP_001189485.1 | 7 - 8 | 1256 | 70 | |
XM_005253231.2 | XP_005253288.1 | 17 - 18 | 2192 | 70 | |
XM_005253232.2 | XP_005253289.1 | 17 - 18 | 2189 | 70 | |
XM_005253233.2 | XP_005253290.1 | 16 - 17 | 2072 | 70 | |
XM_005253234.2 | XP_005253291.1 | 16 - 17 | 2069 | 70 | |
XM_005253235.2 | XP_005253292.1 | 16 - 17 | 2063 | 70 | |
XM_005253238.2 | XP_005253295.1 | 13 - 14 | 1703 | 70 | |
XM_005253239.2 | XP_005253296.1 | 13 - 14 | 1697 | 70 | |
XM_005253240.2 | XP_005253297.1 | 12 - 13 | 1568 | 70 | |
XM_006718388.1 | XP_006718451.1 | 17 - 18 | 2186 | 70 | |
XM_011520482.1 | XP_011518784.1 | 16 - 17 | 1988 | 70 | |
XM_011520483.1 | XP_011518785.1 | 15 - 16 | 1937 | 70 | |
XM_011520484.1 | XP_011518786.1 | 15 - 16 | 1934 | 70 | |
XM_011520485.1 | XP_011518787.1 | 15 - 16 | 1931 | 70 | |
XM_011520486.1 | XP_011518788.1 | 12 - 13 | 1574 | 70 | |
XM_011520488.1 | XP_011518790.1 | 10 - 11 | 1238 | 70 | |
XM_017018583.1 | XP_016874072.1 | 16 - 17 | 2060 | 70 | |
XM_017018584.1 | XP_016874073.1 | 10 - 11 | 1301 | 70 | |
XM_017018585.1 | XP_016874074.1 | 9 - 10 | 1172 | 70 | |
GenBank mRNA | AF098641.1 | - | 3 - 4 | 283 | 70 |
AJ251595.1 | - | 17 - 18 | 2206 | 70 | |
AK129808.1 | - | 7 - 8 | 714 | 70 | |
AK290424.1 | - | 8 - 9 | 885 | 70 | |
AK297776.1 | - | 8 - 9 | 925 | 70 | |
AK304467.1 | - | 7 - 8 | 822 | 70 | |
AK307707.1 | - | 7 - 8 | 780 | 70 | |
AL832642.2 | - | 8 - 9 | 1319 | 70 | |
AY101192.1 | - | 11 - 12 | 1397 | 70 | |
AY101193.1 | - | 8 - 9 | 1002 | 70 | |
BC004372.1 | - | 16 - 17 | 2030 | 70 | |
BC052287.1 | - | 7 - 8 | 873 | 70 | |
BC067348.1 | - | 8 - 9 | 1002 | 70 | |
EF581837.1 | - | 9 - 10 | 1100 | 70 | |
FJ216964.1 | - | 7 - 8 | 822 | 70 | |
M24915.1 | - | 8 - 9 | 1000 | 70 | |
M59040.1 | - | 8 - 9 | 913 | 70 | |
S66400.1 | - | 11 - 12 | 1331 | 70 | |
U40373.1 | - | 8 - 9 | 949 | 70 | |
X55150.1 | - | 11 - 12 | 1404 | 70 | |
X56794.1 | - | 11 - 12 | 1396 | 70 | |
X66733.1 | - | 16 - 17 | 2028 | 70 |