Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FHOD3
- Entrez Gene ID
-
80206
- Gene Name
- formin homology 2 domain containing 3
- Gene Aliases
-
FHOS2, Formactin2
- Chromosome Location
-
Chr.18: 36297696 - 36780220 on Build GRCh38
View Full Product Details