Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018449.3 NP_060919.3 10 - 11 908 65
GenBank mRNA AK026088.1 - 2 - 3 76 65
AK294952.1 - 10 - 11 826 65
AK302335.1 - 8 - 9 729 65
AK302754.1 - 9 - 10 776 65
BC111762.1 - 4 - 5 278 65
BE410205.1 - 7 - 8 596 65

View Full Product Details