Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282529.1 NP_001269458.1 8 - 9 853 69
NM_018449.3 NP_060919.3 12 - 13 1167 69
GenBank mRNA AK026088.1 - 4 - 5 335 69
AK294952.1 - 12 - 13 1085 69
AK299183.1 - 6 - 7 575 69
AK302335.1 - 10 - 11 988 69
AK302754.1 - 11 - 12 1035 69
AK303493.1 - 8 - 9 791 69
BC111762.1 - 6 - 7 537 69

View Full Product Details