The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282529.1 | NP_001269458.1 | 4 - 5 | 470 | 77 |
NM_018449.3 | NP_060919.3 | 5 - 6 | 557 | 77 | |
GenBank mRNA | AK294952.1 | - | 5 - 6 | 475 | 77 |
AK302335.1 | - | 5 - 6 | 537 | 77 | |
AK302754.1 | - | 6 - 7 | 584 | 77 | |
AK303493.1 | - | 4 - 5 | 408 | 77 | |
BG723831.1 | - | 5 - 6 | 564 | 77 |